3-(Butylcarbamoyl)phenylboronic acid ≥97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H52693.MDEA 571 DKK
H52693.MD
3-(Butylcarbamoyl)phenylboronic acid ≥97%
3-(Butylcarbamoyl)phenylboronic acid
Formel: C₁₁H₁₆BNO₃
Molekylvægt: 221,06 g/mol
Smeltepunkt: 216…224 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04115691
CAS-nummer: 397843-70-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR