3-(2,4-Dimethoxyphenyl)-1H-pyrazole ≥98%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H50637.03EA
2190
DKK
H50637.03
3-(2,4-Dimethoxyphenyl)-1H-pyrazole ≥98%
3-(2,4-Dimethoxyphenyl)-1H-pyrazole
Formel:
C₁₁H₁₂N₂O₂ Molekylvægt: 204,23 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02091529 CAS-nummer: 150433-17-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...