Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate ≥98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A13296.06EA 1140 DKK
A13296.06
Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate ≥98%
Ethyl-5-amino-1-phenyl-1H-pyrazole-4-carboxylate
Formel: C₇H₁₁N₃O₂
Molekylvægt: 231,26
Smeltepunkt: 97…102 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00020731
CAS-nummer: 16078-71-0
EINECS: 240-224-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR