Bis(pyridine)iodonium tetrafluoroborate ≥95.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAB2539-1GEA
1420
DKK
TCIAB2539-1G
Bis(pyridine)iodonium tetrafluoroborate ≥95.0% (by titrimetric analysis)
Bis(pyridine)iodonium tetrafluoroborate
Formel:
C₁₀H₁₀BF₄IN₂ Molekylvægt: 371,91 g/mol Smeltepunkt: 151°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD03703393 CAS-nummer: 15656-28-7 UN nummer: 1759 ADR: 8,III |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Iodometric Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...