6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid ≥97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B20289.03EA 567 DKK
B20289.03 B20289.06
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid ≥97%
6-Oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid
Formel: C₅H₆N₂O₃
Smeltepunkt: 196…198 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052030
CAS-nummer: 27372-38-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR