2-Aminoimidazo[1,2-a]pyridine 97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H58726.03EA 4110 DKK
H58726.03
2-Aminoimidazo[1,2-a]pyridine 97%
2-Aminoimidazo[1,2-a]pyridine
Formel: C₇H₇N₃
Smeltepunkt: 114…126 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09836159
CAS-nummer: 39588-26-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR