Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H35393.03EA
2720
DKK
H35393.03
Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₀H₉ClN₂O₂ Molekylvægt: 224,65 g/mol Smeltepunkt: 79…82 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09864797 CAS-nummer: 67625-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...