Ethyl vanillate ≥98.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAV0051-25GEA
641
DKK
TCIAV0051-25G
Ethyl vanillate ≥98.0% (by GC)
Ethyl vanillate
Formel:
C₁₀H₁₂O₄ Molekylvægt: 196,20 g/mol Kogepunkt: 293°C |
MDL Number:
MFCD00017269 CAS-nummer: 617-05-0 EINECS: 210-503-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 48.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...