VWR® Q Path® Histological Fixatives, AFA
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR® Q path®
AFA0150AF59001EA 196 DKK
AFA0150AF59001 AFA0020AF59001 AFA0060AF59001 11656713.
VWR® Q Path® Histological Fixatives, AFA
Farvestoffer Mikroskopi og farvningsreagenser
Alcohol Formalin Acetic acid (AFA) is a ready-to-use fixative for tissue samples.
Each 20 ml pot contains 5 ml. Each 60 ml pot contains 20 ml. Each 150 ml pot contains 60 ml.