Keep my session open?
Ending In 
The session is expired
Din session er udløbet. For din sikkerhed har vi logget dig ud.
Vil du logge ind igen?
OBS! Det er ikke længere muligt at skrive til @dk.vwr.com benyt istedet @vwr.com. Kontakt evt vores kundeservice.
Combination pH electrodes, sensION™ Platinum series
Leverandør: Hach
sensION™
These standard combination pH electrodes feature a replaceable gel reference and a built-in temperature sensor (except for model 5191500). Electrodes come with two electrolyte cartridges and a fixed cable.
  • Replaceable gel reference electrolyte cartridge requires low maintenance and maximises the life of your electrode
  • Accurate and easy to use
  • Rugged and durable: Designed to withstand the rigors of outdoor testing and frequent use
  • Flexible: Suitable for field and laboratory use
The 5191500 model has a flat end that requires minimal immersion and smaller sample volumes, and can be used for surface measurements in water quality applications.
BESTIL
Udvid alle / Skjul alle
Beskrivelse Model pH temp. Materiale Elektrolyt Membran Ø×L Tilslutning Pakn VWR katalog nummer Mængde
Combined pH electrode with built-in temperature sensor sensION Platinum 0 - 14 0...45 °C Epoxy KCl (replaceable) Open junction 12,7×241 mm 5-pin, 0,91 m cable 1 HACH5191000
Combined pH electrode with built-in temperature sensor sensION Platinum 0 - 14 0...45 °C Epoxy KCl (replaceable) Open junction 12,7×241 mm BNC + 3,5 mm phone connector, 0,91 m cable 1 HACH5191011
Combined pH electrode with built-in temperature sensor sensION Platinum 0 - 14 0...45 °C Epoxy KCl (replaceable) Open junction 12,7×241 mm BNC + DIN, 0,91 m cable 1 HACH5191022
Combined pH electrode sensION Platinum, flat end 0 - 13 0...45 °C Epoxy KCl (replaceable) Open junction 12,7×229 mm BNC, 0,91 m cable 1 HACH5191500
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Skriv en bedømmelse
Brug for hjælp?
  • Uanset hvad du mener, positivt eller negativt, er det vigtigt for os.
  • Det tager kun få sekunder at give os feedback.
  • Din feedback vil blive offentliggjort på internettet.
VWR.com vil offentliggøre dit navn som vwr.com bruger.
Du kan bruge dette navn, eller redigere og gemme et kaldenavn.
Vi anbefaler at brugerne bruger deres første og sidste initialer
Bekræft venligst branchen du arbejder i
Eksempel: farmaceutisk
Bekræft venligst dit arbejde
Eksempel: Klinisk assistent
Bekræft venligst Byen
Eksempel: New York
Bedømmelser
Samlet vurdering
Gennemsntilig vurdering
Sortér alle efter:
Enter Details
Select the article for which you are looking for Certificate
Anfør Lotnummeret. Feltet er mandatory. skriv "x" for VWR kemikalier hvis du ikke kender nummeret
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse katalog Nummer Availability Enhed Din pris Price Per Antal