Long swab rinse kits, SRK®
Leverandør: COPAN Diagnostics
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SRK®
710-0431EA 231 DKK
710-0431 710-0472
Long swab rinse kits, SRK®
Podepinde Podepinde med renseopløsning
Swab rinse kits (SRK®) for microbiological examination and quantitative analyses.
  • Swabs are separate from cap
  • Isotonic salt solution to neutralise and deactivate disinfectants and sanitising agents
  • Rinse solution used to moisten the sterile swab
Suitable for sampling and transporting the swab after collection.
Emballering: Rør og applikator indpakket i plastik. Hver æske indeholder 25 poser.