Cryogenic chest freezers
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
THBRULT10140-9-VEA 213450 DKK
THBRULT10140-9-V THBRULT7150-9-M THBRULT10140-9-M THBRULT7150-9-V
Cryogenic chest freezers
Cryogenic storage chest freezers designed for long-term storage of biological samples. Maintain uniform temperatures of −140 and −150 °C (safely below the −130 °C glass transition temperature of water).
  • Provides temperature uniformity and improved cell viability
  • Alarm system triggers audible and visual warning during power failure or temperature deviation
  • Microprocessor control system with touchpad data entry and digital display allows for intuitive operation
It is constructed of heavy-gauge steel with high-impact powder paint finish. It contains heavy-duty counterbalanced lid with integrated keyed lock, double-seal lid gaskets minimise frost buildup and recessed heavy-duty swivel casters.
Refrigeration system comprises of mixed refrigerant autocascade with a single orbital air-cooled compressor for cyrogenic storage, CFC/HCFC free refrigerants maximize cooling and minimize environmental impact and downfeed evaporator for efficient flow of refrigerants, and washable condenser filter keeps free of dust to maintain peak cooling efficiency.
Monitoring system helps to monitor performance and automatically adjust to internal and external conditions through liferguard compressor protection. Voltage boost indicator alerts the low voltage below safe parameters and indicates voltage boost system is activated.
Bestillingsoplysninger: Racks must be ordered separately.