Stand Alone MINI vacuum degassing systems, Systec®
Leverandør: IDEX Health & Science
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Systec® MINI
560-0163EA 34400 DKK
560-0163
Stand Alone MINI vacuum degassing systems, Systec®
Afgassere
The Systec® Stand Alone MINI degassing system is a highly efficient in-line module that removes dissolved gases from fluids. Its unique design ensures reliable and continuous operation and the highest level of continuous performance available without the need for helium degassing. Up to two solvent lines may be degassed simultaneously by one unit. The low internal volume of each AF channel offers very quick equilibration. The module includes a closed loop vacuum control board with validation output.
  • Ultra-high degassing efficiency
  • Easy to prime
  • ZHCR® patented control eliminates baseline fluctuations
  • Excellent chemical compatibility flow path
  • 2-pin vacuum level validation output
  • CE and RoHS compliant
Degassing line path int.Ø: 1,14 mm
Maximum operating pressure: 4,8
Certificeringer: Certified under the following CE testing standards: EN 61326-1; EN 55011; EN 61300-3-2; EN 61300-3-3 and EN 61010-1.
Bestillingsoplysninger: Supplied with AC adapter and four interchangeable plugs: North America/Japan, UK, continental Europe and Australia.