Digital anemometer, DA-100 NT®
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.09192.0001EA 43880 DKK
MERC1.09192.0001
Digital anemometer, DA-100 NT®
Digital anemometer for the calibration of the MAS-100 NT® air samplers (except MAS-100 CG Ex®).
Designed for the precise measurement of air volume (litre per minute), air velocity (meter per second), and air temperature (°C).