4-Fluoro-3-iodobenzoic acid ≥97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H33762.03EA 2170 DKK
H33762.03 H33762.MD
4-Fluoro-3-iodobenzoic acid ≥97%
4-Fluoro-3-iodobenzoic acid
Formel: C₇H₄FIO₂
Smeltepunkt: 173…175 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03788556
CAS-nummer: 403-18-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR