4-(2-Methylphenyl)-3-thiosemicarbazide ≥98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L09382.06EA 536 DKK
L09382.06 L09382.14
4-(2-Methylphenyl)-3-thiosemicarbazide ≥98%
4-(2-Methylphenyl)-3-thiosemicarbazide
Formel: C₈H₁₁N₃S
Smeltepunkt: 145…148 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041277
CAS-nummer: 614-10-8
ADR: 6.1,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR