4'-Isothiocyanatoacetophenone ≥98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L11927.06EA 1520 DKK
L11927.06 L11927.03
4'-Isothiocyanatoacetophenone ≥98%
4'-Isothiocyanatoacetophenone
Formel: C₉H₇NOS
Kogepunkt: 112 °C (6 mmHg)
Smeltepunkt: 77…81 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041100
CAS-nummer: 2131-57-9
EINECS: 000-000-0
ADR: 8,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR