2-Chloro-4-(trifluoromethyl)benzyl alcohol ≥97%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H33376.03EA 3140 DKK
H33376.03 H33376.MD
2-Chloro-4-(trifluoromethyl)benzyl alcohol ≥97%
2-Chloro-4-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆ClF₃O
Smeltepunkt: 50…52 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04972757
CAS-nummer: 56456-51-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR