3-Bromo-5-(trifluoromethoxy)benzyl alcohol ≥97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H31815.06EA
3070
DKK
H31815.06
H31815.03
3-Bromo-5-(trifluoromethoxy)benzyl alcohol ≥97%
3-Bromo-5-(trifluoromethoxy)benzyl alcohol
Formel:
C₈H₆BrF₃O₂ Molekylvægt: 271,03 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09999512 CAS-nummer: 1026201-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...