4-(Methylsulphonyl)-2-nitrobenzoic acid

Leverandør: EHRENSTORFER GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

EHERC15143800EA 1840 DKK
EHERC15143800
4-(Methylsulphonyl)-2-nitrobenzoic acid
4-(Methylsulphonyl)-2-nitrobenzoic acid
Formel: C₈H₇NO₆S
Molekylvægt: 245,2100 g/mol
MDL Number: MFCD01941299
CAS-nummer: 110964-79-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR