Infusion valves, Ultrasite®
Leverandør: B. Braun
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BRAU415111EA 5190 DKK
BRAU415111
Infusion valves, Ultrasite®
Infusioner
Simplify IV technique by direct insertion into a closed system.
  • Non-hemolytic and lipid resistant
  • Prevents needlestick injuries
  • Reduces catheter occlusions
Compatible with Luer lock and Luer slip, valve opens and closes automatically with each Luer taper.