2,5-Dimethylcyclohexanol (mixture of isomers) ≥99.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAD0704-25GEA
690
DKK
TCIAD0704-25G
2,5-Dimethylcyclohexanol (mixture of isomers) ≥99.0% (by GC)
2,5-Dimethylcyclohexanol (mixture of isomers)
Formel:
C₈H₁₆O Kogepunkt: 179°C Flammepunkt: 69°C |
MDL Number:
MFCD00001504 CAS-nummer: 3809-32-3 EINECS: 223-281-3 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...