6-Methoxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAM2256-5GEA
493
DKK
TCIAM2256-5G
6-Methoxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
6-Methoxy-2-naphthaleneboronic acid
Formel:
C₁₁H₁₁BO₃ Molekylvægt: 202,02 g/mol Smeltepunkt: 311°C |
MDL Number:
MFCD03093087 CAS-nummer: 156641-98-4 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...