Calorimeter, C 1
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
IKAC10002412EA 144800 DKK
IKAC10002412 IKAC10002414 IKAC10002415 IKAC10002417
Calorimeter, C 1
The oxygen bomb calorimeter C1 possesses a high degree of automation. It operates with an isoperibol static jacket and a light attachable combustion chamber.
  • Automatic oxygen filling, venting and flushing
  • Operates either with a chiller (KV 600) or tap water when using the water heater C1.20
  • Interfaces for PC (USB-B), printer (serial interface), balance (serial interface)
  • Automatic ignition with fixed ignition wire as well as ignition energy determination for each experiment
  • Automatic water filling and draining
  • Traditional heavy screw threaded bomb has been replaced by a light combustion chamber
It can be operated through a user panel and with the Calwin C 6040 software. This software allows for additional features in data handling with Microsoft SQL, LIMS and correction calculations.
Leveringsinformation: Calorimeter is supplied with power supply unit, inflow conduit, return conduit, discharge hose, round cable clamp, connection tube, venting hose, mains cable, O-rings, C 1.104 water bath additive, C 723 Benzoic acid, C 1.1012 Organizer, O-ring-grease Molykote 55, handle and operating instructions. The packages additionally include a chiller or a heater.