trans-2-Heptenyl acetate ≥97.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAA1145-10MLEA
1110
DKK
TCIAA1145-10ML
trans-2-Heptenyl acetate ≥97.0% (by GC)
trans-2-Heptenyl acetate
Formel:
C₉H₁₆O₂ Kogepunkt: 193°C |
MDL Number:
MFCD00036584 CAS-nummer: 16939-73-4 EINECS: 241-002-3 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...