6-Heptenyl acetate ≥97.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAH0849-5MLEA
1450
DKK
TCIAH0849-5ML
6-Heptenyl acetate ≥97.0% (by GC)
6-Heptenyl acetate
Formel:
C₉H₁₆O₂ Kogepunkt: 74°C |
MDL Number:
MFCD00191656 CAS-nummer: 5048-30-6 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...