(S)-(-)-1-Benzyl-3-acetamidopyrrolidine 98%, ee 99%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

L19689.06EA 1480 DKK
L19689.06
(S)-(-)-1-Benzyl-3-acetamidopyrrolidine 98%, ee 99%
(S)-(-)-1-Benzyl-3-acetamidopyrrolidine
Formel: C₁₃H₁₈N₂O
Smeltepunkt: 44…46 °C
MDL Number: MFCD03788746
CAS-nummer: 114636-30-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR