Isobornyl acetate til syntese, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel


M-Clarity™ quality level = MQ200

8.14998.0250EA 301 DKK
8.14998.0250 8.14998.9999
Isobornyl acetate til syntese, Sigma-Aldrich®
Isobornyl acetate
Formel: C₁₂H₂₀O₂
Molekylvægt: 196,29 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00135943
CAS-nummer: 125-12-2
EINECS: 204-727-6

Bestil nu

Specifikationer Testresultater

Solubility 0.16 g/l (20 °C)
Molar Mass 196.29 g/mol
Boiling Point 215 °C (1013 hPa)
Vapor Pressure 0.13 hPa (20 °C)
Flash Point 88 °C
Density 0.98 g/cm3 (20 °C)
Ignition Point 430 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR