Ethyl isodehydroacetate 95%
Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ACRO373900250EA 797 DKK
ACRO373900250
Ethyl isodehydroacetate 95%
Ethyl isodehydroacetate
Formel: C₁₀H₁₂O₄
Molekylvægt: 196,1999 g/mol
Kogepunkt: 290…295 °C (760 mmHg)
Smeltepunkt: 17…18 °C
Massefylde: 1,167 g/cm³
Flammepunkt: >110 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006643
CAS-nummer: 3385-34-0
EINECS: 222-195-3
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5135 to 1.5165 (20°C, 589 nm)
GC ≥94 %