1H-Benzotriazole-1-methanol ≥98.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB1903-25GEA
583
DKK
TCIAB1903-25G
1H-Benzotriazole-1-methanol ≥98.0% (by titrimetric analysis)
1H-Benzotriazole-1-methanol
Formel:
C₇H₇N₃O Molekylvægt: 149,1500 g/mol |
MDL Number:
MFCD00179118 CAS-nummer: 28539-02-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 148.0 to 151.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...