(R)-(+)-3-Acetamidopyrrolidine ≥98.0% (by titrimetric analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAA1169-5GEA 2630 DKK
TCIAA1169-5G
(R)-(+)-3-Acetamidopyrrolidine ≥98.0% (by titrimetric analysis)
(R)-(+)-3-Acetamidopyrrolidine
Formel: C₆H₁₂N₂O
Molekylvægt: 128,17 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00082729
CAS-nummer: 131900-62-4
https://dk.vwr.com/stibo/web/std.lang.all/90/17/28929017.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 55.0 to 60.0 deg-C
Specific rotation [a]20/D +45.0 to +49.0 deg (C=1, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR