D-(+)-Glucose-[6,6-D₂] (99% D)

Leverandør: CAMBRIDGE ISOTOPE LABS., INC.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

CAMBDLM-349-10EA 9070 DKK
CAMBDLM-349-10
D-(+)-Glucose-[6,6-D₂] (99% D)
D-(+)-Glucose-[6,6-D₂]
Formel: C₆H₁₀D₂O₆
MDL Number: MFCD00064367
CAS-nummer: 18991-62-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR