Ginkgolide J, Supelco®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


M-Clarity™ quality level = MQ100

89556-5MGEA 3280 DKK
89556-5MG
Ginkgolide J, Supelco®
Ginkgolide J
Formel: C₂₀H₂₄O₁₀
CAS-nummer: 107438-79-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR