4'-Methoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA1307-500GEA
813
DKK
TCIAA1307-500G
TCIAA1307-25G
4'-Methoxyacetoacetanilide ≥98.0% (by HPLC, total nitrogen)
4'-Methoxyacetoacetanilide
Formel:
C₁₁H₁₃NO₃ Molekylvægt: 207,2300 g/mol |
MDL Number:
MFCD00008783 CAS-nummer: 5437-98-9 EINECS: 226-615-6 |
Specifikationer Testresultater
Appearance | White to Blue powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 115.0 to 118.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...