Dodecacarbonyltriiron ≥96% (dry weight) stabilised
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Stabilization:
stabiliseret med methanol
13059.09EA
1450
DKK
13059.09
Dodecacarbonyltriiron ≥96% (dry weight) stabilised
Dodecacarbonyltriiron
Reducing agent (reduces NO2), catalyst for alkyne trimerization.
Formel:
C₁₂Fe₃O₁₂ Molekylvægt: 503,66 g/mol Smeltepunkt: 140 °C (dec.) Massefylde: 2,00 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00135617 CAS-nummer: 17685-52-8 EINECS: 241-668-5 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...