Single channel pipettes, mechanical, variable volume, PIPETMAN®
Leverandør: Gilson
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PIPETMAN®
GILSF144067CGEA 2890 DKK
GILSF144067CG GILSF144066CG 613-7160 613-6191 GILSF144054MCG GILSF144067C GILSF144066C 613-5946 GILSF144054MC 613-5945 613-5889 GILSF144055MC 613-5944 613-5949 613-5948 613-5947 613-6501 613-6500 613-5950
Single channel pipettes, mechanical, variable volume, PIPETMAN®
Pipetter Automatiske pipetter
Mechanical air-displacement pipette which offers robustness, accuracy and precision as well as a very low pipetting force for the highest comfort.
  • Very low pipetting forces thanks to easy glide piston displacement
  • Colour-coded push button ensures easy identification and GLP compliance
  • Made of PVDF and stainless steel
  • Equipped with clip ejector for convenient access to the serviceable parts
  • Blue ring positioned under the thumbwheel allows to identify PIPETMAN® and to distinguish it from the Classic and Neo models
Pipettes are individually calibrated and delivered with a certificate of conformity. Each pipette bears an engraved identification number for traceability.
Bestillingsoplysninger: Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories. Please note: PIPETMAN P5000 and P10 ml do not have tip ejectors.