Pallets and sumps for storage of hazardous materials
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MSGA18444EA 6660 DKK
MSGA18444
Pallets and sumps for storage of hazardous materials
Galvanized steel construction with painted catchment sum. Safe storage of pollutant and flammable liquids.
  • Compact dimensions, perfect for the storage of 205 L drums
  • With louvres at the front doors
  • Suitable for internal or external applications
  • Lockable
  • Maximum load: 700 kg/m² (when the load is evenly distributed across the storage area)
  • Sump capacity: 205 L
Leveringsinformation: Delivered in dismantled state for self-assembly.