Pallets and sumps for IBC station
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MSGA2684EA 11460 DKK
MSGA2684 MSGA8437 MSGA8833 MSGA2683
Pallets and sumps for IBC station
PE or galvanised grid.
  • Corrosion resistance
  • Maximum loading capacity 2000 kg
For the safe storage of water pollutants.