Du Søgte efter: Kompetente celler
Kompetente celler er celler som har fået modificeret/manipuleret deres cellevæg således at det er lettere at indføre fremmed DNA ind i cellerne. Celler bliver kompetente ved en process, som benytter kalcium klorid og varmechok, og kompetente celler vokser meget hurtigere og bliver gjort kompetente for hurtigere vækst end celler i andre stadier.
Kompetente celler er celler som har fået modificeret/manipuleret deres cellevæg således at det er lettere at indføre fremmed DNA ind i cellerne. Celler bliver kompetente ved en process, som benytter kalcium klorid og varmechok, og kompetente celler vokser meget hurtigere og bliver gjort kompetente for hurtigere vækst end celler i andre stadier.
25  results were found
SearchResultCount:"25"
Sort Results
Listevisning Easy View (new)
Tilfreds med din søgning? - rate den her.
Varenummer: (AGLS200129)
Leverandør: AGILENT
Beskrivelse: XL-1 Red Competent Cells are ideal for introducing random mutations into a cloned gene of interest.
UOM: 1 * 1 mL
Leverandør: AGILENT
Beskrivelse: XL10-Gold ultracompetent cells transform large plasmids and ligated DNA with highest efficiency.
Leverandør: AGILENT
Beskrivelse: SURE 2 competent cells are ideal for routine cloning of DNA with secondary structures.
Leverandør: AGILENT
Beskrivelse: BJ5183 electroporation competent cells are a recombination proficient bacterial strain.
Leverandør: AGILENT
Beskrivelse: Tackle two common bacterial gene expression hurdles: protein insolubility and codon bias.
Leverandør: AGILENT
Beskrivelse: BL21 competent cells are an all-purpose strain for high-level protein expression and easy induction.
Leverandør: AGILENT
Beskrivelse: JM Competent Cells are recombination- and E. coli K12 restriction-proficient.
Leverandør: AGILENT
Beskrivelse: High efficiency competent cells in convenient packaging designed to satisfy higher throughput requirements.
Leverandør: AGILENT
Beskrivelse: XL1 and XL2-Blue Competent Cells are versatile for routine cloning and allow blue-white colour screening for high efficiency cloning of methylated DNA...
Varenummer: (AGLS200123)
Leverandør: AGILENT
Beskrivelse: Phage display libraries from electroporation-competent cells with high transformation efficiency.
UOM: 1 * 5 Items
Varenummer: (AGLS200134)
Leverandør: AGILENT
Beskrivelse: Easily induce and express phosphorylated proteins, which can be used to screen expression libraries, to affinity purify or blot proteins that interact...
UOM: 1 * 1 mL
Varenummer: (AGLS200231)
Leverandør: AGILENT
Beskrivelse: SCSI supercompetent and DH5α competent cells are classic E. coli strains engineered for competence.
UOM: 1 * 1 mL
Varenummer: (AGLS200159)
Leverandør: AGILENT
Beskrivelse: Achieve the highest transformation efficiencies available when cloning large plasmids and ligated DNA.
UOM: 1 * 5 Items
Varenummer: (AGLS230246)
Leverandør: AGILENT
Beskrivelse: Protein Expression competent cell packs efficiently test multiple strains for expression.
UOM: 1 * 1 kt
Varenummer: (AGLS200247)
Leverandør: AGILENT
Beskrivelse: SCS110 competent cells for generating unmethylated DNA improve quality and yield of miniprep DNA.
UOM: 1 * 1 mL
Varenummer: (AGLS200172)
Leverandør: AGILENT
Beskrivelse: ABLE Competent Cells for toxic clones reduce copy number four to ten fold compared to the XL1-Blue strain.
UOM: 1 * 1 mL
Ring og spørg om pris
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Lager for denne post er begrænset, men kan være til rådighed på et lager tæt på dig. Sørg for, at du er logget ind på webstedet, så tilgængelige bestand kan vises. Hvis call vises stadig, og du har brug for hjælp, bedes du ringe til os på - 43 86 87 88.
Dette produkt er mærket som begrænset og kan kun købes af godkendte Shipping konti. Hvis du har brug for yderligere hjælp, e-mail VWR Regulatory Department på Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dette produkt er blevet blokeret for salg via hjemmesiden. Kontakt kundeservice for mere information.
Det originale produkt er ikke længere tilgængelig. Den viste erstatning er tilgængelig.
Produkt (er) markeret med dette symbol er udgået - sælges indtil lageret er tomt. Alternativer kan være tilgængelige ved at søge med VWR-katalognummeret ovenfor. Hvis du har brug for yderligere hjælp, bedes du kontakte kundeservice på telefon 4386 8788
1 - 16 of 25
no targeter for Bottom