Barrels, wide neck, with lid
Leverandør: Semadeni
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SEMA2830EA 999 DKK
SEMA2830 SEMA2831 SEMA2832 SEMA2833 SEMA0726 SEMA0725
Barrels, wide neck, with lid
Tønder, fustager, trommer
Made of HDPE, blue, round (except model 2831 which is rectangular).
  • With handles
  • Good chemical and temperature resistance
  • With UN approval
Heavy duty open top drums with PE lid and galvanised clamping ring. Suitable for storage, disposal and transport of laboratory materials.
The 150 L barrel is specially designed as an ISO container.
Certificeringer: 30, 60, 120 and 220 L barrels are approved for food use.