3'-Benzyloxyacetophenon
Leverandør: LGC Standards PROMOCHEM
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PROOMM0035.50EA
13170
DKK
PROOMM0035.50
3'-Benzyloxyacetophenon
3'-Benzyloxyacetophenon
Formel:
C₁₅H₁₄O₂ Molekylvægt: 226,27 g/mol |
MDL Number:
MFCD00026221 CAS-nummer: 34068-01-4 EINECS: 251-816-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...