1-Acetyl-2-naphthol ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAH0936-25GEA 427 DKK
TCIAH0936-25G
1-Acetyl-2-naphthol ≥98.0% (by GC)
1-Acetyl-2-naphthol
Formel: C₁₂H₁₀O₂
Molekylvægt: 186,21 g/mol
Kogepunkt: 178°C
Smeltepunkt: 64 °C
MDL Number: MFCD00012131
CAS-nummer: 574-19-6
EINECS: 209-369-4
https://dk.vwr.com/stibo/web/std.lang.all/09/54/28970954.jpg

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 66.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR