4'-(2-Chloroethyl)acetophenone tech. 90%, Technical Grade
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
L13800.06EA
1110
DKK
L13800.06
4'-(2-Chloroethyl)acetophenone tech. 90%, Technical Grade
4'-(2-Chloroethyl)acetophenone
Formel:
C₁₀H₁₁ClO Molekylvægt: 182,65 Kogepunkt: 112 °C (0,1 mmHg) Massefylde: 1,150 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00016350 CAS-nummer: 69614-95-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...