(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAD3677-5GEA
2000
DKK
TCIAD3677-5G
TCIAD3677-1G
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-2,2-Dimethylcyclopropanecarboxylic acid
Formel:
C₆H₁₀O₂ Kogepunkt: 100°C |
MDL Number:
MFCD08460918 CAS-nummer: 14590-53-5 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +122.0 to +136.0 deg (C=1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...