(R)-(+)-Pulegone
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ROTH5168.1EA
0
DKK
ROTH5168.1
(R)-(+)-Pulegone
(R)-(+)-Pulegone
Formel:
C₁₀H₁₆O Molekylvægt: 152,23 g/mol |
MDL Number:
MFCD00063000 CAS-nummer: 89-82-7 EINECS: 201-943-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...