1-Chloro-7-phenylheptane ≥98%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
L08817.03EA
1370
DKK
L08817.03
1-Chloro-7-phenylheptane ≥98%
1-Chloro-7-phenylheptane
Formel:
C₁₃H₁₉Cl Molekylvægt: 210,75 g/mol Kogepunkt: 94…96 °C (0,3 mmHg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00039392 CAS-nummer: 71434-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...