Nonadecanophenone ≥97.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAN0664-25GEA
1330
DKK
TCIAN0664-25G
Nonadecanophenone ≥97.0% (by GC)
Nonadecanophenone
Formel:
C₂₅H₄₂O |
MDL Number:
MFCD00094242 CAS-nummer: 103044-68-4 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 63.0 to 66.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...