Hexadecanophenone ≥95.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAH0590-5GEA
386
DKK
TCIAH0590-5G
TCIAH0590-25G
Hexadecanophenone ≥95.0% (by GC)
Hexadecanophenone
Formel:
C₂₂H₃₆O Kogepunkt: 248°C |
MDL Number:
MFCD00008997 CAS-nummer: 6697-12-7 |
![]() |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 58.0 to 61.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...