4-Amino-3-bromobenzoic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA1969-5GEA
214
DKK
TCIAA1969-5G
4-Amino-3-bromobenzoic acid ≥98.0% (by titrimetric analysis)
4-Amino-3-bromobenzoic acid
Formel:
C₇H₆BrNO₂ |
MDL Number:
MFCD03407439 CAS-nummer: 6311-37-1 |
![]() |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 94.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 210.0 to 214.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...