(3-Carboxypropyl)triphenylphosphonium bromide ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAC1635-25GEA
452
DKK
TCIAC1635-25G
(3-Carboxypropyl)triphenylphosphonium bromide ≥95.0% (by HPLC, titration analysis)
(3-Carboxypropyl)triphenylphosphonium bromide
Formel:
C₂₂H₂₂BrO₂P Molekylvægt: 429,30 g/mol |
MDL Number:
MFCD00274196 CAS-nummer: 17857-14-6 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 246.0 to 250.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...